Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stachydrine Hydrochloride, TCI America™
SDP

Catalog No. S035810MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S035810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S035810MG Supplier TCI America Supplier No. S035810MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4136-37-2
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD00060174
InChI Key DUNMULOWUUIQIL-RGMNGODLSA-N
PubChem CID 44150282
IUPAC Name (2S)-1,1-dimethylpyrrolidin-1-ium-2-carboxylic acid;chloride
SMILES C[N+]1(CCCC1C(=O)O)C.[Cl-]

Specifications

Quantity 10 mg
Formula Weight 179.64
Physical Form Crystalline Powder
Chemical Name or Material Stachydrine Hydrochloride
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.