Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stachyose tetrahydrate | 10094-58-3 | 98.1% | 738.64 | 10 MG
SDP

Catalog No. 5000437162
Encompass_Preferred

Stachyose tetrahydrate is a natural saccharide, a tetrasaccharide composed of two α-D-galactose units, one D-glucose unit, and one D-fructose unit. This product appears as a white to yellow solid and is tested to comply with stringent specifications, ensuring high quality for various research applications.

  • High purity: 98.10% by HPLC
  • Appearance: White to yellow solid
  • Recommended storage conditions: 4°C, sealed, away from moisture and light
  • Long-term solvent storage: -80°C for 6 months or -20°C for 1 month in sealed, moisture and light-protected conditions
  • Quality tested: Complies with specified standards

Catalog No. 50-004-37162 Supplier Medchemexpress LLC Supplier No. HY11352910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stachyose tetrahydrate is a natural saccharide, a tetrasaccharide composed of two α-D-galactose units, one D-glucose unit, and one D-fructose unit. This product appears as a white to yellow solid and is tested to comply with stringent specifications, ensuring high quality for various research applications.

  • High purity: 98.10% by HPLC
  • Appearance: White to yellow solid
  • Recommended storage conditions: 4°C, sealed, away from moisture and light
  • Long-term solvent storage: -80°C for 6 months or -20°C for 1 month in sealed, moisture and light-protected conditions
  • Quality tested: Complies with specified standards

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.