Learn More
Cayman Chemical Stachyose (hydrate), Purity: ≥95%, CAS: 54261-98-2, 5 g
SDP
Catalog No. 502950093
Encompass_Preferred
An oligosaccharide and prebiotic; reduces the number of colonic lesions and serum TNF-α, IL-6, IL-10, and IL-17a levels, as well as increases gut levels of Akkermansia and Lactobacillus, in a rat model of DSS-induced colitis at 1 g/kg per day; decreases serum LPS, IL-6, and TNF-α levels and enriches Phascolarctobacterium, Bilophila, Oscillospira, and Turicibacter in the gut microbiota in a rat model of STZ- and high-fat diet-induced diabetes; enhances berberine-induced blood glucose control and improvements in insulin resistance, as well as decreases the fecal concentration of short-chain fatty acids, in the KKAγ mouse model of type 2 diabetes
Catalog No. 50-295-0093 Supplier Cayman Chemical Supplier No. 343225 G
An oligosaccharide and prebiotic; reduces the number of colonic lesions and serum TNF-α, IL-6, IL-10, and IL-17a levels, as well as increases gut levels of Akkermansia and Lactobacillus, in a rat model of DSS-induced colitis at 1 g/kg per day; decreases serum LPS, IL-6, and TNF-α levels and enriches Phascolarctobacterium, Bilophila, Oscillospira, and Turicibacter in the gut microbiota in a rat model of STZ- and high-fat diet-induced diabetes; enhances berberine-induced blood glucose control and improvements in insulin resistance, as well as decreases the fecal concentration of short-chain fatty acids, in the KKAγ mouse model of type 2 diabetes