Learn More
Quantum Storage Systems QUANTUM STORAGE SYSTEMS Hang and Stack Bin: 11 in x 18 1/2 in
Catalog No. NC3246461
Encompass_Preferred
QUANTUM STORAGE SYSTEMS Hang and Stack Bin: 11 in x 18 1/2 in x 10 1/4 in, Finish: Chrome; Model QMB560C. 5 bins per carton.Inside Dimensions: 18" x 10-5/8" x 10"Heavy-duty wire mesh hanging and stacking bins are woven or welded from wire and have small openings that allow clear visibility of contents and let air, liquids, and dust pass through. Does not include Dividers, Optional side hangers (sold separately) allow bins stack one on top of the other.
Catalog No. NC3246461 Supplier Quantum Storage Systems Supplier No. QMB560CCS
QUANTUM STORAGE SYSTEMS Hang and Stack Bin: 11 in x 18 1/2 in x 10 1/4 in, Finish: Chrome; Model QMB560C. 5 bins per carton.Inside Dimensions: 18" x 10-5/8" x 10"Heavy-duty wire mesh hanging and stacking bins are woven or welded from wire and have small openings that allow clear visibility of contents and let air, liquids, and dust pass through. Does not include Dividers, Optional side hangers (sold separately) allow bins stack one on top of the other.