Learn More
MSC MFG Tray Tote, Composite, 300 lb Capacity, White, 1 ea
Catalog No. 5000513541
Encompass
This fiberglass composite tote is designed for heavy-duty industrial storage, offering a robust 300 lb load capacity. Its durable construction ensures the container will not bend, sag, dent, or corrode under load, making it suitable for demanding environments including machine integration and robotic applications. The tote features a smooth surface and radial edges for safe handling and maintains structural integrity across a wide temperature range.
  • Constructed from glass-reinforced composite for high durability
  • Impervious to cutting oils, greases, solvents, detergents, mild acids, and alkaline solutions
  • Operating temperature range of -60°F to 250°F
  • Smooth outside surfaces and radial edges allow for safe handling
  • Compatible with optional lids and dollies to simplify transport and protection
Catalog No. 50-005-13541 Supplier MSC Supplier No. 00343822
This fiberglass composite tote is designed for heavy-duty industrial storage, offering a robust 300 lb load capacity. Its durable construction ensures the container will not bend, sag, dent, or corrode under load, making it suitable for demanding environments including machine integration and robotic applications. The tote features a smooth surface and radial edges for safe handling and maintains structural integrity across a wide temperature range.
  • Constructed from glass-reinforced composite for high durability
  • Impervious to cutting oils, greases, solvents, detergents, mild acids, and alkaline solutions
  • Operating temperature range of -60°F to 250°F
  • Smooth outside surfaces and radial edges allow for safe handling
  • Compatible with optional lids and dollies to simplify transport and protection