Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Stack-Rack Space-Saver Tips: 1-200 μL

Catalog No. 02707106 Shop All Fisherbrand Products
Change view
Click to view available options
Format:
Rack, 5 x 200
Rack, 5 x 96
Color:
Natural
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Color
02-707-106 Rack, 5 x 200 Yellow
02-707-105 Rack, 5 x 200 Natural
02-707-115 Rack, 5 x 96 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-707-106

Increase limited bench space with Stack Racks

  • Racks nest together with each rack serving as a lid for the rack below
  • Rack packs contain five racks of tips, with a base and dust cover
  • Available in 5 x 96 or 5 x 200 formats
  • Certified by independent laboratory
  • Nonsterile; RNase, DNase, DNA, and pyrogen free
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Yellow
Autoclavable Autoclavable
Length (English) 2 in.
Length (Metric) 5 cm
Clean Claims DNase, RNase, DNA, and Pyrogen Free
Format Rack, 5 x 200
Tip Style Beveled
For Use With (Equipment) Universal Fit
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.