Learn More
OHAUS Stack Tray MultiPlate Genie
Catalog No. 01804381 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Tray
Catalog No. Product Type
01-804-381 Tray
1 options
Catalog No. 01-804-381 Supplier OHAUS Supplier No. 83041324
Only null left
Add to Cart
Add to Cart
Optional Stacking Tray for use on MultiPlate Genie (models SI-4100 - SI-410X).
  • Provides for increased capacity up to 8 microplates
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tray