Learn More
JDM LAB SOLUTIONS LLC Stackable Mini Roller
Catalog No. NC2385848
Encompass
Variable speed and LED display allow for precise controlAnti-corrosive and wear-resistant construction is safe for high humidity and CO2 environmentsEasily add or remove rollers to fit a wide range of tube and bottle sizesStackable design allows for efficient use of benchtop surface areaIncluded wired remote externally control instrument when running inside the incubator
Catalog No. NC2385848 Supplier JDM LAB SOLUTIONS LLC Supplier No. MR01UA
Variable speed and LED display allow for precise controlAnti-corrosive and wear-resistant construction is safe for high humidity and CO2 environmentsEasily add or remove rollers to fit a wide range of tube and bottle sizesStackable design allows for efficient use of benchtop surface areaIncluded wired remote externally control instrument when running inside the incubator