Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kord™ 85 x 13 Ultra Plate Stackable Slippable Petri Dishes
SCI_ED

Catalog No. S43570
Change view
Click to view available options
Diameter (Metric):
85 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
S43570 85 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43570 Supplier Kord™ Supplier No. 2950
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For standard microbiology and general laboratory use.

  • Slippable rimmed lids provide semi-stacking feature
  • Deep dish design in mono style
  • Polystyrene construction
  • With ISO mark target

Specifications

Closure Type Slippable
Description New Ultra Plate
Material Polystyrene
Shape Round
Sterility Sterile
Type Petri Dish
Well Count 1
No. per Pack 30/Sleeve
Size 85 x 13 mm
Height (Metric) 13 mm
Diameter (Metric) 85 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.