Learn More
Medicus Health Stackable Specimen Cup Rack
SDP
Catalog No. NC0981245
Encompass
∙Stackable Specimen Cup Racks are designed to hold specimen cups for histology and urinalysis ∙Racks hold 12 specimen cups up to 2.25" in Diameter∙Guaranteed to fit your specimen cups for histology, microbiology and urinalysis departments∙Patent Pending handle stacks racks in place when empty, partially full and filled∙Stacking design keeps specimens organized and upright∙Great for organizing, storing and transporting pap smear collection vials and cups∙Space saving design conserves refrigerator space, counter space and storage space∙Perfect for histology specimens and urine specimens∙Can be used for transporting specimens from collection areas∙Epoxy coated wire rack is autoclavable and safe for use in freezers, refrigerators and incubators∙Dimensions: 10"L x 8"W x 4.25"H
Catalog No. NC0981245 Supplier Medicus Health Supplier No. 5524M1
∙Stackable Specimen Cup Racks are designed to hold specimen cups for histology and urinalysis ∙Racks hold 12 specimen cups up to 2.25" in Diameter∙Guaranteed to fit your specimen cups for histology, microbiology and urinalysis departments∙Patent Pending handle stacks racks in place when empty, partially full and filled∙Stacking design keeps specimens organized and upright∙Great for organizing, storing and transporting pap smear collection vials and cups∙Space saving design conserves refrigerator space, counter space and storage space∙Perfect for histology specimens and urine specimens∙Can be used for transporting specimens from collection areas∙Epoxy coated wire rack is autoclavable and safe for use in freezers, refrigerators and incubators∙Dimensions: 10"L x 8"W x 4.25"H