Learn More
OHAUS Stackable Tray Roto-Shake Genie
Catalog No. 01804421 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-421 Shaker Attachment
1 options
Catalog No. 01-804-421 Supplier OHAUS Supplier No. 83041368
Add to Cart
Add to Cart
A tall non-skid metal tray.
  • This tray can be stacked onto the Roto-Shake Genie's existing tray to rock various containers including T-flasks, petri dishes, boxes, and sealed bags, increasing the capacity for rocking by double-stacking two trays.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment