Learn More
Analytical Sales & Services Stackable Tray with SILANIZED 8x30 Shell Vials
Catalog No. 5000806519
Encompass
Stackable Tray Loaded with SILANIZED 8x30 Shell Vials (84001-CASE)\nfor Photoredox/Optimization Plates (96-well)\n\n• Recommended for top performance - our thorough QC process ensures less evaporation compared to loose vials\n• Makes loading and transferring your vials easier!\n• Can be used with Powder Transfer Plate (96T105) to mix/transfer powders between vials\n• Can also be used as a stand-alone collection/sampling device\n\nIncludes Tray and 96 SILANIZED Vials
Catalog No. 50-008-06519 Supplier Analytical Sales & Services Supplier No. 884001S
Stackable Tray Loaded with SILANIZED 8x30 Shell Vials (84001-CASE)\nfor Photoredox/Optimization Plates (96-well)\n\n• Recommended for top performance - our thorough QC process ensures less evaporation compared to loose vials\n• Makes loading and transferring your vials easier!\n• Can be used with Powder Transfer Plate (96T105) to mix/transfer powders between vials\n• Can also be used as a stand-alone collection/sampling device\n\nIncludes Tray and 96 SILANIZED Vials