Learn More
OHAUS Stackable Wire Rack Enviro-G
Catalog No. 01804355 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-355 Shaker Attachment
1 options
Catalog No. 01-804-355 Supplier OHAUS Supplier No. 83041376
Add to Cart
Add to Cart
Stackable Wire Rack may be used in the Enviro-Genie and the Incubator-Genie.
  • It enables to increase shelf space and also as a temperature controlled chamber.
  • Two wire racks will fit inside the chamber to provide 3 levels for containers.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment