Learn More
Med Vet International Stacked Locking Hole Plate 2.4mm 10 hole 73mm Long, J1221F
Catalog No. 5000580962
Encompass_Preferred
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 10 Hole 73mm long
Catalog No. 50-005-80962 Supplier Med Vet International Supplier No. JOR1221F
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 10 Hole 73mm long