Learn More
Med Vet International Stacked Locking Hole Plate 2.4mm 11 hole 80mm Long, J1221G
Catalog No. 5000580935
Encompass_Preferred
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 11 Hole 80mm long
Catalog No. 50-005-80935 Supplier Med Vet International Supplier No. JOR1221G
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 11 Hole 80mm long