Learn More
Med Vet International Stacked Locking Hole Plate 2.4mm 12 hole 87mm Long, J1221H
Catalog No. 5000581194
Encompass_Preferred
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 12 Hole 87mm long
Catalog No. 50-005-81194 Supplier Med Vet International Supplier No. JOR1221H
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 12 Hole 87mm long