Learn More
Med Vet International Stacked Locking Hole Plate 2.4mm 13 hole 94mm Long, J1221I
Catalog No. 5000581280
Encompass_Preferred
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 13 Hole 94mm long
Catalog No. 50-005-81280 Supplier Med Vet International Supplier No. JOR1221I
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 13 Hole 94mm long