Learn More
Med Vet International Stacked Locking Hole Plate 2.4mm 14 hole 101mm Long, J1221J
Catalog No. 5000581305
Encompass_Preferred
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 14 Hole 101mm long
Catalog No. 50-005-81305 Supplier Med Vet International Supplier No. JOR1221J
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 14 Hole 101mm long