Learn More
Med Vet International Stacked Locking Hole Plate 2.4mm 15 hole 108mm Long, J1221K
Catalog No. 5000581195
Encompass_Preferred
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 15 Hole 108mm long
Catalog No. 50-005-81195 Supplier Med Vet International Supplier No. JOR1221K
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 15 Hole 108mm long