Learn More
Med Vet International Stacked Locking Hole Plate 2.4mm 16 hole 115mm Long, J1221L
Catalog No. 5000581279
Encompass_Preferred
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 16 Hole 115mm long
Catalog No. 50-005-81279 Supplier Med Vet International Supplier No. JOR1221L
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 16 Hole 115mm long