Learn More
Med Vet International Stacked Locking Hole Plate 2.4mm 17 hole 122mm Long, J1221M
Catalog No. 5000581133
Encompass_Preferred
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 17 Hole 122mm long
Catalog No. 50-005-81133 Supplier Med Vet International Supplier No. JOR1221M
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 17 Hole 122mm long