Learn More
Med Vet International Stacked Locking Hole Plate 2.4mm 6 Hole 45mm Long, J1221B
Catalog No. 5000581134
Encompass_Preferred
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 6 Hole 45mm long
Catalog No. 50-005-81134 Supplier Med Vet International Supplier No. JOR1221B
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 6 Hole 45mm long