Learn More
Med Vet International Stacked Locking Hole Plate 2.4mm 7 hole 52mm long, J1221C
Catalog No. 5000581306
Encompass_Preferred
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 7 Hole 52mm long
Catalog No. 50-005-81306 Supplier Med Vet International Supplier No. JOR1221C
Jorvet 2.4 locking plates have stacked locking holes as is the case with our 2.7 plates. The twin start hole in the plate is the same size as the 2.7 screws, cortical 2.4, cortical 2.7, and cancellous 3.0. Hole spacing is 7mm.2.4mm Locking Hole Plate 7 Hole 52mm long