Learn More
BINDER™ Stacking Adapter
SDP
Catalog No. 502875640 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB 65
KB/KB PRO 130, KBF/KBF PRO 130
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5640 KB/KB PRO 130, KBF/KBF PRO 130
50-287-5638 KB 65
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-287-5640 Supplier BINDER™ Supplier No. 90510048
Add to Cart
Add to Cart
Stacking adapter to stack two devices.
  • Used to safely stack two identical devices
  • For use with KB 65, KB 130, KB PRO 130, KBF 130, or KBF PRO 130 models
Specifications
For Use With (Equipment) KB/KB PRO 130, KBF/KBF PRO 130
Material Plastic
Type Stacking Adapter