Learn More
Kuhner Shaker Inc STACKING ADAPTER FOR LS-Z..TO
Catalog No. 502569941
Encompass
Stacking adapter for LS-Z..To stack two LS-Z/Kelvin+..Limited shaking frequency for..The upper shaker:..Up to 11kg laod:..12.5/25/50mm: 300RPM..11-25kg load:..12.5mm: 200RPM..25mm: 200RPM..50mm: 170RPM..
Catalog No. 50-256-9941 Supplier Kuhner Shaker Inc Supplier No. SMZ104911
Stacking adapter for LS-Z..To stack two LS-Z/Kelvin+..Limited shaking frequency for..The upper shaker:..Up to 11kg laod:..12.5/25/50mm: 300RPM..11-25kg load:..12.5mm: 200RPM..25mm: 200RPM..50mm: 170RPM..