Learn More
Bioworld Stacking Gel Buffer (4X), 50 mL
Catalog No. 50196666
Encompass_Preferred
Product DescriptionStacking gel buffer has a concentration of 4X and contains 0.5M Tris at a pH value of 6.8 and 0.4% SDS. Final concentrations of Tris and SDS after dilution is 0.125M and 0.1% respectively. Product is stored at room temperature.
Catalog No. 50-196-666 Supplier Bioworld Supplier No. 105300091
Product DescriptionStacking gel buffer has a concentration of 4X and contains 0.5M Tris at a pH value of 6.8 and 0.4% SDS. Final concentrations of Tris and SDS after dilution is 0.125M and 0.1% respectively. Product is stored at room temperature.