Learn More
Research Products International Corp Stacking PCR Rack, 5 per Package
SDP
Catalog No. 50998182
Encompass
Innovative rack provides a convenient workstation for PCR applications or can be used as a durable storage rack for all 0.2ml PCR tubes, strip tubes or 96-well plates. Racks are molded from heavy duty polypropylene for use at temperatures from -80°C to +121°C. Supplied five per case in assorted bright colors that include green, yellow, blue, violet and red.
Catalog No. 50-998-182 Supplier Research Products International Corp Supplier No. 248635
Innovative rack provides a convenient workstation for PCR applications or can be used as a durable storage rack for all 0.2ml PCR tubes, strip tubes or 96-well plates. Racks are molded from heavy duty polypropylene for use at temperatures from -80°C to +121°C. Supplied five per case in assorted bright colors that include green, yellow, blue, violet and red.