Learn More
Thomas Scientific Stacking Platform 12" X 12" w / Dimpled Mat
SDP
Catalog No. 501461133
Encompass
The BenchBlotter™ is specifically optimized with ideal rocking speed and tilt angle for blot washing, gel staining/destaining and hybridization applications. Available with an optional dual platform accessory, the BenchBlotter™ offers a sizeable work space, while maintaining a compact footprint and low profile design. Lightweight and easy to carry, it can be easily transported from the lab bench to a cold room or incubator. The large load capacity (7 lbs.), optional dimpled mat for centrifuge tubes and multiple platform tie-down points make this rocker suitable for a wide variety of general purpose applications.
Catalog No. 50-146-1133 Supplier Thomas Scientific Supplier No. 1227U64
The BenchBlotter™ is specifically optimized with ideal rocking speed and tilt angle for blot washing, gel staining/destaining and hybridization applications. Available with an optional dual platform accessory, the BenchBlotter™ offers a sizeable work space, while maintaining a compact footprint and low profile design. Lightweight and easy to carry, it can be easily transported from the lab bench to a cold room or incubator. The large load capacity (7 lbs.), optional dimpled mat for centrifuge tubes and multiple platform tie-down points make this rocker suitable for a wide variety of general purpose applications.