Learn More
Fritsch Milling And Sizing Inc Stacking Ring for 125-150Ml Premium Line
SDP
Catalog No. 501534594
Encompass
Stacking ring for all grinding bowls for Planetary Mill PULVERISETTE 5 premium line 125-150 ml volume.
Essential, if 2 grinding bowls 125-150 ml volume per grinding bowl holder are used.
Catalog No. 50-153-4594 Supplier Fritsch Milling And Sizing Inc Supplier No. 50.6830.00
Stacking ring for all grinding bowls for Planetary Mill PULVERISETTE 5 premium line 125-150 ml volume.
Essential, if 2 grinding bowls 125-150 ml volume per grinding bowl holder are used.