Learn More
USA Scientific Inc Stacking Tray with Dimpled Mat
Catalog No. 502744748
Encompass
Stacking Tray for 3D Rocker, gyratory rocker, large 12"x12" with dimpled mat  (3.0" separation)
Catalog No. 50-274-4748 Supplier USA Scientific Inc Supplier No. 25231073
Stacking Tray for 3D Rocker, gyratory rocker, large 12"x12" with dimpled mat  (3.0" separation)