Learn More
Enterprise Technology Solutions Staedtler Engineers Pro Triangular Scale, Mars, ALM, 12", Silver
SDP
Catalog No. 5000739447
Encompass_Preferred
Professional-quality Engineer's triangular scale features a metal construction using solid-extruded aluminum. Solid-extruded aluminum is resistant to all weather conditions. Color-coded grooves allow an accurate selection of the desired scale. Easy-to-read imprinted graduations are designed to hold up against fading.
Catalog No. 50-007-39447 Supplier Enterprise Technology Solutions Supplier No. STD987M1834BK
Professional-quality Engineer's triangular scale features a metal construction using solid-extruded aluminum. Solid-extruded aluminum is resistant to all weather conditions. Color-coded grooves allow an accurate selection of the desired scale. Easy-to-read imprinted graduations are designed to hold up against fading.