Learn More
Enterprise Technology Solutions Staedtler Markers, Erasable on Film, Fine Point, 10/BX, Black
SDP
Catalog No. 5000827167
Encompass_Preferred
Nonpermanent marker is designed for overhead projection writing. Quick-drying, water-based, nonpermanent ink won't bleed or smudge. Write on overhead transparency film or such glossy surfaces as photos, transparency sleeves, slide frames, cromalins, glass, acrylic and vinyl. Water-soluble ink can be wiped off film using a damp cloth. It is great for crafts, graphic arts, photography, lab work or print shop. Fiber-tip pen features a polypropylene barrel and cap with a clip to attach to notebooks, pockets and more.
Catalog No. 50-008-27167 Supplier Enterprise Technology Solutions Supplier No. STD3169
Nonpermanent marker is designed for overhead projection writing. Quick-drying, water-based, nonpermanent ink won't bleed or smudge. Write on overhead transparency film or such glossy surfaces as photos, transparency sleeves, slide frames, cromalins, glass, acrylic and vinyl. Water-soluble ink can be wiped off film using a damp cloth. It is great for crafts, graphic arts, photography, lab work or print shop. Fiber-tip pen features a polypropylene barrel and cap with a clip to attach to notebooks, pockets and more.