Learn More
Micro Biologics Inc 3P STAFF EPIDERMUS QUICK STIX
SDP
Catalog No. NC0501959
Encompass_Preferred
Staphylococcus epidermidis derived from ATCC® 12228™* Two KWIK-STIK™ units at E4. For use with Cepheid® GeneXpert System only
Catalog No. NC0501959 Supplier Micro Biologics Inc 3P Supplier No. 0371MSSE
Staphylococcus epidermidis derived from ATCC® 12228™* Two KWIK-STIK™ units at E4. For use with Cepheid® GeneXpert System only