Learn More
Selleck Chemical LLC Stafia-1-E0485-5MG
SDP
Supplier:  Selleck Chemical LLC E04855MG
Encompass_Preferred
Stafia-1 is the first STAT5a inhibitor that inhibits STAT5a (IC50 22 2 M Ki 10 9 M) with at least 9 fold selectivity over STAT5b and higher selectivity against other STAT family members
Catalog No. 50-238-9622