Learn More
Medchemexpress LLC HY-136546 5mg Medchemexpress, Stafia-1 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1365465MG
Encompass_Preferred
Medchemexpress, HY-136546 5mg Stafia-1 CAS: Stafia-1 is a potent STAT5a inhibitor (K i=10.9 μM, IC50=22.2 μM). Stafia-1 displays high selectivity over STAT5b and other STAT family members. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1446