Learn More
Electron Microscopy Sciences Stage Micrometer S20, Double micrometer scale Scale, Length
SDP
Supplier:  Electron Microscopy Sciences 6804320
Encompass_Preferred
Stage graticules are used for the routine calibration of eyepiece patterns, particularly when alternating between objectives on one microscope or when using the same graticule in different microscopes. Their robust construction makes them ideal for student use and for instructional purposes. The scale or grid is centered on a glass disc mounted in a black anodized aluminum slide 75 mm x 24 mm x 2 mm thick.
Scale: Double micrometer scale
Divisions: 2 mm in 0.01 mm and 0.1"in 0.005"
Catalog No. 50-284-77