Learn More
Electron Microscopy Sciences Stage Graticules (Micrometers) S78
SDP
Supplier:  Electron Microscopy Sciences 6804378
Encompass_Preferred
For reflected light the scale is etched through highly reflective vacuum coated metal. When viewed under vertical illumination the scale appears black against a bright background. Micrometer scale is 1 mm with 0.01 mm divisions.
Catalog No. 50-312-26