Learn More
Electron Microscopy Sciences EMS Stage Micrometer, SM-1
SDP
Supplier:  Electron Microscopy Sciences 602101PGC
Encompass_Preferred
Chrome on Glass. Linear Reference Scales: Glass size - 2" x 5.25". Scale Length - 5 inch X-axis only, 125 mm X-axis only. Divisions - English = 0.01”; Metric = 100um (0.1 mm). With numerical labeling every 1 mm and 0.1” With Calibration.
Catalog No. 50-340-08