Learn More
Electron Microscopy Sciences EMS Microscope Stage Micrometer SM-12
SDP
Supplier:  Electron Microscopy Sciences 6021012PGC
Encompass_Preferred
SM-12 is useful for calibration of magnification in microscopes and other optical magnification systems.. It comes in a sturdy wooden box. Chrome on Glass. Linear Reference Scales: Glass Size - 1" x 3" (25 mm x 75 mm). Scale Length - 0.08 inch X axis, 2.0 mm X & Y axis. Divisions- English = 0.001”; Metric = 25um (0.025 mm). No numerical scale labeling. With Calibration.
Catalog No. 50-340-12