Learn More
Electron Microscopy Sciences EMS Microscope Stage Micrometer SM-3
SDP
Supplier:  Electron Microscopy Sciences 602103PGC
Encompass_Preferred
SM-3 is useful for calibration of magnification in microscopes and other optical magnification systems. Its auto-location features can be used for testing of an image analysis auto-location system. Chrome on Glass. Linear Reference Scales: Glass Size - 1" x 3". Scale Length - 0.04 inch X & Y axis, 10.0 mm X & Y axis. Divisions- English = 0.0005”; Metric = 10um. With numerical labeling every 0.1 mm and 0.05”. With Calibration.
Catalog No. 50-340-18