Learn More
Electron Microscopy Sciences EMS Microscope Stage Micrometer SM-5
SDP
Supplier:  Electron Microscopy Sciences 602105PG
Encompass_Preferred
Chrome on Glass. Linear Reference Scales: Glass Size - 1.5" x 3". Scale Length - 1 inch X & Y axis, 25 mm X-axis only, Divisions- English = 0.001”; Metric = 25um (0.025 mm), With numerical labeling every 1 mm and 0.1”. All EMS micrometers come in a sturdy wooden box.
Catalog No. 50-340-17