Learn More
Thermo Scientific STAGETIP C18 MAT 200UL 96PC
Supplier:  Thermo Scientific SP301
Encompass
StageTips, C18 material, 200a�L tip. For rapid desalting and step elution of peptide mixtures. Loading capacity in 10 i1/4g range. Includes 2.5 mL Eppendorf CombiTip. 96 pcs.
Catalog No. NC0028383