Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Stahl Aerobic Oxidation Tempo Solution

Catalog No. 79654925ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-654-925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 79-654-925ML Supplier Sigma Aldrich Supplier No. 79654925ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Stahl Aerobic Oxidation TEMPO solution contains 1-methylimidazole (NMI), 2,2'-bipyridyl (bpy), and TEMPO. It is widely employed for the oxidation of alcohols.This convenience solution can be used in tandem with tetrakisacetonitrile copper(I) triflate (685038) for the oxidation of primary alcohols through aerobic conditions developed by the Stahl Research Group.Technology Spotlight: Aerobic Alcohol Oxidation Solutions Stable TEMPO and ABNO Catalyst Solutions for User-Friendly (bpy)Cu/Nitroxyl- Catalyzed Aerobic Alcohol OxidationPractical Aerobic Alcohol Oxidation with Cu/Nitroxyl and Nitroxyl/NOx Catalyst Systems

Specifications

CAS 1329422-59-4
Quantity 25 mL
Synonym (S)-2-Amino-4-(7-hydroxy-2-oxo-2H-chromen-4-yl)butanoic acid; Schultz Fluorescent UAA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.