Learn More
Med Vet International Stain #1, J0326ASD1, 120mL, Each
Catalog No. 5000583116
Encompass_Preferred
A very useful, specialized stain. Use for identification of: Johnes disease, cryptosporidium, acid fast organisms. 126ml bottle.
Catalog No. 50-005-83116 Supplier Med Vet International Supplier No. JOR326ASD1
A very useful, specialized stain. Use for identification of: Johnes disease, cryptosporidium, acid fast organisms. 126ml bottle.