Learn More
Med Vet International Stain #2, J0326ASD2, 120mL, Each
Catalog No. 5000583061
Encompass_Preferred
A very useful, specialized stain. Use for identification of: Johnes disease, cryptosporidium, acid fast organisms. 126ml bottle.
Catalog No. 50-005-83061 Supplier Med Vet International Supplier No. JOR326ASD2
A very useful, specialized stain. Use for identification of: Johnes disease, cryptosporidium, acid fast organisms. 126ml bottle.