Learn More
AdipoGen Stain 3-I
SDP
Catalog No. 502560658
Encompass_Preferred
Chemical. CAS 155862-91-2. Formula C18H16BrN3O2S. MW 418.31. Synthetic. Class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient Epsilon∼ >20,000 in EtOH.
Catalog No. 50-256-0658 Supplier AdipoGen Supplier No. CDXI0032M050
Chemical. CAS 155862-91-2. Formula C18H16BrN3O2S. MW 418.31. Synthetic. Class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient Epsilon∼ >20,000 in EtOH.