Learn More
Med Vet International Stain #3, J0326ASD3, 120mL, Each
Catalog No. 5000583005
Encompass_Preferred
A very useful, specialized stain. Use for identification of: Johnes disease, cryptosporidium, acid fast organisms. 126ml bottle.
Catalog No. 50-005-83005 Supplier Med Vet International Supplier No. JOR326ASD3
A very useful, specialized stain. Use for identification of: Johnes disease, cryptosporidium, acid fast organisms. 126ml bottle.