Learn More
AdipoGen Stain 7-I
SDP
Catalog No. 502560094
Encompass_Preferred
Chemical. CAS 155862-95-6. Formula C27H22BrN3O6. MW 564.38. Synthetic. New class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance.
Catalog No. 50-256-0094 Supplier AdipoGen Supplier No. CDXS0030M250
Chemical. CAS 155862-95-6. Formula C27H22BrN3O6. MW 564.38. Synthetic. New class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance.