Learn More
AdipoGen Stain 7-II
SDP
Catalog No. 502560091
Encompass_Preferred
Chemical. CAS 155863-03-9. Formula C29H24BrN3O6. MW 590.42. Synthetic. New class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance.
Catalog No. 50-256-0091 Supplier AdipoGen Supplier No. CDXS0031M250
Chemical. CAS 155863-03-9. Formula C29H24BrN3O6. MW 590.42. Synthetic. New class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance.