Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen Stain Away™ Wipes

Catalog No. 23730626 Shop All Scigen Inc Products
Change view
Click to view available options
Quantity:
60 Pk., 240/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-730-626 60 Pk., 240/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-730-626 Supplier Scigen Supplier No. 4275
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stain removing wipes removes biological stains and dyes from hands and surfaces.

  • Used on hands and non-porous surfaces
  • Stain-absorbing wipes instantly remove biological dyes and stains, literally lifting them off your skin
  • Perfect for removing Cytology, Hematology, and Histology stains

Specifications

Sterility Non-sterile
Appearance White
Container Type Pop-Up Wipe Dispenser
Content And Storage Store at Room Temperature
Description Stain removing wipes
Dimensions (L x W) 5 x 8 in.
For Use With (Application) Removing biological stains and dyes from hands
Includes Instructions for Use
Quantity 60 Pk., 240/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.